Tisk…
Hydroxid hlinitý, fine powder
Dodavatel: Merck
Synonyms: Hydrargillite, Aluminum hydroxide
1.01093.1000 1.01093.9025
1.01093.1000EA 0 CZK
1.01093.1000 1.01093.9025
Hydroxid hlinitý, fine powder
Aluminium hydroxide
Vzorec: Al(OH)₃
MW: 78 g/mol
Hustota: 2,42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003420
CAS číslo: 21645-51-2
EINECS: 244-492-7
Merck Index: 13,00342