Hydroxid hlinitý ≥76.5%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Hydrargillite, Aluminum hydroxide
12366.22
12366.36
12366.A3
12366.22EA
1010
CZK
12366.22
12366.36
12366.A3
Hydroxid hlinitý ≥76.5%
Aluminium hydroxide
Purity: 76.5% min
CAS No.: 21645-51-2
Molecular Formula: AlH3O3
Formula Weight: 78.00
Physical Form: Granular
Appearance: White
Solubility: Practically insoluble in water. Soluble in alkaline aqueous solution, HCl, H2SO4 solution
Merck Reference: 14,342
MDL No.: MFCD00003420
As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmetics
Vzorec:
Al(OH)₃ MW: 78 g/mol Bod tání: 300 °C Hustota: 2,42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003420 CAS číslo: 21645-51-2 EINECS: 244-492-7 Merck Index: 14,342 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...