Tripropyl borate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Purity: 98%
CAS No.: 688-71-1
Molecular Formula: C9H21BO3
Formula Weight: 188.08
Flash Point: 32°(90°F)
Physical Form: Liquid
Density: 0.861
Refractive Index: 1.3950
Sensitivity: Moisture Sensitive
Appearance: Colourless
Boiling Point: 175-177°
Solubility: Soluble in organic solvents. Decomposes in water
UN No.: 3272
MDL No.: MFCD00009366
Vzorec:
C₉H₂₁BO₃ Bod varu: 175…177 °C Hustota: 0,861 Bod vzplanutí: 32 °C (90 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009366 CAS číslo: 688-71-1 EINECS: 211-703-9 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...