Tisk…

Triethyl borate ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A18728.0F A18728.AE A18728.AP
A18728.0FEA 18570 CZK
A18728.0F A18728.AE A18728.AP
Triethyl borate ≥97%
Triethyl borate

Purity: 97% CAS No.: 150-46-9 Molecular Formula: C6H15BO3 Formula Weight: 145.99 Flash Point: 11°(51°F) Refractive Index: 1.3740 Odour: Mild Melting Point: -85° Boiling Point: 117-118° Beilstein Reference: 1699052 UN No.: 1176 MDL No.: MFCD00009073

Vzorec: C₆H₁₅BO₃
MW: 145,99 g/mol
Bod varu: 117…118 °C
Bod tání: –85 °C
Hustota: 0,858
Bod vzplanutí: 11 °C (51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012846
CAS číslo: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR