Tributyl borate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Tri-n-butylborate
A19322.0F
A19322.AE
A19322.AP
A19322.0FEA
10020
CZK
A19322.0F
A19322.AE
A19322.AP
Tributyl borate ≥98%
Tributyl borate
Purity: 98%
CAS No.: 688-74-4
Molecular Formula: [CH3(CH2)3O]3B
Formula Weight: 230.16
Flash Point: 93°(199°F)
Refractive Index: 1.4095
Melting Point:
Vzorec:
C₁₂H₂₇BO₃ MW: 230,15 g/mol Bod varu: 230…235 °C Bod tání: < –70 °C Hustota: 0,853 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009434 CAS číslo: 688-74-4 EINECS: 211-706-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...