Triisopropyl borate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tri-i-propylborate
M-Clarity™ quality level = MQ200
197335-750G
197335-80G
SIAL197335-750GEA
6210
CZK
SIAL197335-750G
SIAL197335-80G
Triisopropyl borate, Sigma-Aldrich®
Triisopropyl borate
Vzorec:
C₉H₂₁BO₃ MW: 188,07 g/mol Bod varu: 139 °C (1013 hPa) Bod tání: –59 °C Hustota: 0,82 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008872 CAS číslo: 5419-55-6 EINECS: 226-529-9 UN: 2616 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...