Tisk…

Triisopropyl borate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Tri-i-propylborate

A17592.0F A17592.AE A17592.AP
A17592.0FEA 25190 CZK
A17592.0F A17592.AE A17592.AP
Triisopropyl borate ≥98%
Triisopropyl borate

Purity: 98+% CAS No.: 5419-55-6 Molecular Formula: C9H21BO3 Formula Weight: 188.08 Flash Point: 17°(62°F) Refractive Index: 1.3760 Odour: Alcohol-like Melting Point: -59° Boiling Point: 139-141° Beilstein Reference: 1701469 UN No.: 2616 MDL No.: MFCD00008872

Vzorec: C₉H₂₁BO₃
MW: 188,07 g/mol
Bod varu: 139…141 °C
Bod tání: –59 °C
Hustota: 0,815
Bod vzplanutí: 17 °C (62 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS číslo: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR