Oxid inditý, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Indium trioxide, Diindium trioxide
M-Clarity™ quality level = MQ100
289418-10G
289418-50G
SIAL289418-10GEA
4450
CZK
SIAL289418-10G
SIAL289418-50G
Oxid inditý, Sigma-Aldrich®
Oxid inditý
Vzorec:
In₂O₃ MW: 277,64 g/mol Bod tání: 715,05 °C Hustota: 7,18 g/cm³ (20 °C) |
MDL Number:
MFCD00011060 CAS číslo: 1312-43-2 EINECS: 215-193-9 Merck Index: 12,04984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...