Aluminium oxide, γ-α-phase ≥99.98% (metals basis)
Dodavatel: Thermo Scientific
10459.14
10459.22
10459.36
10459.14EA
1520
CZK
10459.14
10459.22
10459.36
Aluminium oxide, γ-α-phase ≥99.98% (metals basis)
Aluminium oxide, γ-α-phase
Purity: 99.98%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 0.01-0.02 Micron Powder, S.A. 100m2/g
Density: 3.965
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Vzorec:
Al₂O₃ Bod varu: 2980 °C Bod tání: 2045 °C Hustota: 3,965 Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...