Tisk…

Aluminium oxide, α-phase ≥99.95% (metals basis)

Dodavatel: Thermo Fisher Scientific

Synonyms: ɑ-Tonerde, α-Alumina

42573.22 42573.36 42573.A3
42573.22EA 1350 CZK
42573.22 42573.36 42573.A3
Aluminium oxide, α-phase ≥99.95% (metals basis)
Aluminium oxide, α-phase

Purity: 99.95% min CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 0.25-0.45 Micron APS Powder, S.A. 7.2-8.8m2/g Refractive Index: 1.765 Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424

Vzorec: Al₂O₃
MW: 101.96
Bod varu: 2980 °C
Bod tání: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS číslo: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR