Aluminium oxide, α-phase ≥99.99% (metals basis)
Dodavatel: Thermo Scientific
36341.14
36341.22
36341.14EA
3020
CZK
36341.14
36341.22
Aluminium oxide, α-phase ≥99.99% (metals basis)
Aluminium oxide, α-phase
Purity: 99.99%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 2-12mm Pieces
Refractive Index: 1.765
Odour: Odourless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Vzorec:
Al₂O₃ Bod varu: 2980 °C Bod tání: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...