Oxid hlinitý substrate 10x10x0.5 mm, polished both sides A plane
Dodavatel: Thermo Fisher Scientific
Synonyms:
Aluminium(III) oxide, Alumina C, Aluminum oxide
41426.KT
41426.KU
41426.KTEA
3120
CZK
41426.KT
41426.KU
Oxid hlinitý substrate 10x10x0.5 mm, polished both sides A plane
Oxid hlinitý
Molecular Formula: Al2O3
MDL No.: MFCD00003424
Vzorec:
Al₂O₃ MW: 101.96 Bod varu: 2980 Bod tání: 2045 Hustota: 3.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...