Oxid hlinitý ≥99.995% (metals basis), Puratronic®
Dodavatel: Thermo Scientific
10627.14
10627.22
10627.36
10627.14EA
5700
CZK
10627.14
10627.22
10627.36
Oxid hlinitý ≥99.995% (metals basis), Puratronic®
Aluminium oxide
Purity: 99.995%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: Powder
Density: 3.965
Refractive Index: 1.768
Odour: Odourless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Vzorec:
Al₂O₃ Bod varu: 2980 °C Bod tání: 2045 °C Hustota: 3,965 Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 14,356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...