Tisk…

Nitrid boritý

Dodavatel: Thermo Fisher Scientific
Varování

40607.04 40607.09
40607.04EA 4230 CZK
40607.04 40607.09
Nitrid boritý
Nitrid boritý

CAS No.: 10043-11-5 Molecular Formula: BN (cubic) Formula Weight: 24.82 Physical Form: 4-8 Micron Powder Density: 3.48 Odour: Odourless Merck Reference: 14,1346 MDL No.: MFCD00011317

Vzorec: BN
Bod tání: 2527 °C
Hustota: 3,48
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS číslo: 10043-11-5
EINECS: 233-136-6
Merck Index: 14,1346

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR