Tisk…

Indium(III) iodide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

413666-1G 413666-5G
SIAL413666-1GEA 2370 CZK
SIAL413666-1G SIAL413666-5G
Indium(III) iodide, Sigma-Aldrich®
Indium(III) iodide
Vzorec: I₃In
Bod varu: 500 °C (1013 hPa)
Hustota: 4,69 g/cm³ (20 °C)
MDL Number: MFCD00016151
CAS číslo: 13510-35-5
EINECS: 236-839-6
UN: 3260
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR