Aluminium iodide, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Aluminum iodide
M-Clarity™ quality level = MQ100
409324-1G
409324-5G
SIAL409324-1GEA
2580
CZK
SIAL409324-1G
SIAL409324-5G
Aluminium iodide, Sigma-Aldrich®
Aluminium iodide
Vzorec:
AlI₃ MW: 407.7 Bod varu: 382 Bod tání: 191 Hustota: 3.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003427 CAS číslo: 7784-23-8 EINECS: 232-054-8 UN: 3260 ADR: 8 II Merck Index: 12,00358 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...