Aluminium fluoride, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Aluminum fluoride
M-Clarity™ quality level = MQ100
449628-10G
449628-50G
SIAL449628-10GEA
7210
CZK
SIAL449628-10G
SIAL449628-50G
Aluminium fluoride, Sigma-Aldrich®
Aluminium fluoride
Vzorec:
AlF₃ MW: 83,98 g/mol Bod tání: 1040 °C Hustota: 2,88 g/cm³ (20 °C) |
MDL Number:
MFCD00003426 CAS číslo: 7784-18-1 EINECS: 232-051-1 UN: 3260 ADR: 8,II Merck Index: 12,00351 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...