Tisk…

Aluminium fluoride, Patinal®

Dodavatel: Merck
Varování

Synonyms: Aluminum fluoride

1.01600.0100
1.01600.0100EA 0 CZK
1.01600.0100
Aluminium fluoride, Patinal®
Aluminium fluoride
Vzorec: AlF₃
MW: 83,98 g/mol
Bod tání: 1040 °C
Hustota: 2,88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003426
CAS číslo: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8,II
Merck Index: 12,00351

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR