Tisk…

Indium(III) chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Indium Trichloride Chloride, Indium trichloride

M-Clarity™ quality level = MQ200

203440-10G 203440-1G 203440-50G
SIAL203440-10GEA 6190 CZK
SIAL203440-10G SIAL203440-1G SIAL203440-50G
Indium(III) chloride, Sigma-Aldrich®
Indium(III) chloride
Vzorec: Cl₃In
MW: 221.18
Bod varu: 800
Bod tání: 586
Hustota: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011058
CAS číslo: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8 II
Merck Index: 13,04980

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR