Indium(I) chloride ≥99.999% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
10707.06
10707.14
10707.06EA
10020
CZK
10707.06
10707.14
Indium(I) chloride ≥99.999% (metals basis), Puratronic®
Indium(I) chloride
Purity: 99.999%
CAS No.: 13465-10-6
Molecular Formula: InCl
Formula Weight: 150.27
Physical Form: Crystalline Lump
Density: 4.19
Melting Point: ca 225°
Boiling Point: 608°
UN No.: 3260
MDL No.: MFCD00148875
Vzorec:
InCl Bod varu: 608 °C Bod tání: ∼225 °C Hustota: 4,19 Storage Temperature: Ambient |
MDL Number:
MFCD00148875 CAS číslo: 13465-10-6 EINECS: 236-693-3 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...