Tisk…

Gallium(III) chloride, ultra dry ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Gallium trichloride

Ampuled under argon

45026.22 45026.14 45026.06
45026.22EA 25330 CZK
45026.22 45026.14 45026.06
Gallium(III) chloride, ultra dry ≥99.999% (metals basis)
Gallium(III) chloride

Purity: 99.999% CAS No.: 13450-90-3 Molecular Formula: GaCl3 Formula Weight: 176.03 Physical Form: 8mm and down Granules, Ampouled under argon Density: 2.47 Appearance: White Melting Point: 78° Boiling Point: 201° Merck Reference: 14,4348 UN No.: 3260 MDL No.: MFCD00011018

Vzorec: Cl₃Ga
MW: 176.08
Bod varu: 201 °C
Bod tání: 78 °C
Hustota: 2.47
Storage Temperature: Ambient
MDL Number: MFCD00011018
CAS číslo: 13450-90-3
EINECS: 236-610-0
UN: 3260
ADR: 8 II
Merck Index: 14,4348

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR