Tisk…

Indium(III) bromide, anhydrous ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Indium tribromide

40126.09 40126.18
40126.09EA 6630 CZK
40126.09 40126.18
Indium(III) bromide, anhydrous ≥99.99% (metals basis)
Indium(III) bromide

Purity: 99.99% CAS No.: 13465-09-3 Molecular Formula: Br3In Formula Weight: 354.55 Physical Form: Crystalline powder Density: 4.74 Appearance: White to yellow Odour: Odourless Melting Point: ca 436° MDL No.: MFCD00011057

Vzorec: Br₃In
MW: 354.53
Bod varu: 656
Bod tání: ∼ 436 °C
Hustota: 4.74
Storage Temperature: Ambient
MDL Number: MFCD00011057
CAS číslo: 13465-09-3
EINECS: 236-692-8
UN: 3260
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR