Boron tribromide, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
419508-100G
419508-500G
SIAL419508-100GEA
3560
CZK
SIAL419508-100G
SIAL419508-500G
Boron tribromide, Sigma-Aldrich®
Boron tribromide
Vzorec:
BBr₃ MW: 250.52 Bod varu: 91.2 Bod tání: –46 °C Hustota: 2.645 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011312 CAS číslo: 10294-33-4 EINECS: 233-657-9 UN: 2692 ADR: 8 I Merck Index: 13,01337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...