Tisk…

Boron tribromide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

202207-100G 202207-25G
SIAL202207-100GEA 8570 CZK
SIAL202207-100G SIAL202207-25G
Boron tribromide, Sigma-Aldrich®
Boron tribromide
Vzorec: BBr₃
MW: 250,52 g/mol
Bod varu: 91,2 °C (1013 hPa)
Bod tání: –46 °C
Hustota: 2,645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS číslo: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8,I
Merck Index: 13,01337

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR