Hydroxid hořečnatý 95-100.5%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
12367.36
12367.22
12367.A4
12367.36EA
1610
CZK
12367.36
12367.22
12367.A4
Hydroxid hořečnatý 95-100.5%
Hydroxid hořečnatý
Purity: 95-100.5%
CAS No.: 1309-42-8
Molecular Formula: Mg(OH)2
Formula Weight: 58.33
Physical Form: Powder
Density: 2.36
Appearance: White
Odour: Odourless
Melting Point: 360°dec
Solubility: Soluble in dilute acids. Practically insoluble in water
Merck Reference: 14,5670
MDL No.: MFCD00011104
Flame retardant in plastics, such as PVC, as it produces less toxic fumes
Vzorec:
Mg(OH)₂ Hustota: 2,36 Storage Temperature: Ambient |
MDL Number:
MFCD00011104 CAS číslo: 1309-42-8 EINECS: 215-170-3 Merck Index: 14,5670 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...