Tisk…

Barium hydroxide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

433373-1KG 433373-250G
SIAL433373-1KGEA 3700 CZK
SIAL433373-1KG SIAL433373-250G
Barium hydroxide, Sigma-Aldrich®
Barium hydroxide
Vzorec: H₂BaO₂
MW: 171,34 g/mol
Bod tání: 325 °C
Hustota: 2,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003443
CAS číslo: 17194-00-2
EINECS: 241-234-5
UN: 3262
ADR: 8,III
Merck Index: 12,01006

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR