Tisk…

Oxid hořečnatý heavy ≥96%

Dodavatel: Thermo Fisher Scientific

Synonyms: Magnesia usta

44237.22 44237.36 44237.A1
44237.22EA 2300 CZK
44237.22 44237.36 44237.A1
Oxid hořečnatý heavy ≥96%
Oxid hořečnatý

Purity: 96% CAS No.: 1309-48-4 Molecular Formula: MgO Formula Weight: 40.30 Physical Form: -325 Mesh Powder Density: 3.58 Refractive Index: 1.736 Odour: Odourless Melting Point: 2852° Boiling Point: 3600° Merck Reference: 14,5677 MDL No.: MFCD00011109

Vzorec: MgO
MW: 40.3
Bod varu: 3600 °C
Bod tání: 2852 °C
Hustota: 3.58
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS číslo: 1309-48-4
EINECS: 215-171-9
Merck Index: 14,5677

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR