Tisk…

Barium manganate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Barium manganate(VI)

M-Clarity™ quality level = MQ100

210196-100G 210196-25G 210196-5G
SIAL210196-100GEA 17310 CZK
SIAL210196-100G SIAL210196-25G SIAL210196-5G
Barium manganate, Sigma-Aldrich®
Barium manganate
Vzorec: BaMnO₄
MW: 256,27 g/mol
Hustota: 5,2 g/cm³ (20 °C)
MDL Number: MFCD00011514
CAS číslo: 7787-35-1
EINECS: 232-109-6
UN: 3087
ADR: 5.1,III
Merck Index: 13,00984

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR