Magnesium fluoride hydrate (≥87% MgF₂)
Dodavatel: Thermo Scientific
|
Varování
|
A12830.0E
A12830.36
A12830.0EEA
4500
CZK
A12830.0E
A12830.36
Magnesium fluoride hydrate (≥87% MgF₂)
Magnesium fluoride hydrate
CAS No.: 7783-40-6
Molecular Formula: MgF2.xH2O
Formula Weight: 62.30(anhy)
Odour: Odourless
Merck Reference: 14,5665
MDL No.: MFCD00011108
Vzorec:
F₂Mg·H₂O MW: 62,30 g/mol (anhy) Bod tání: 1266 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS číslo: 7783-40-6 EINECS: 231-995-1 Merck Index: 14,5665 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...