Tisk…

Fluorid hořečnatý ≥99.9% (metals basis excluding Ca, Na) (max. 1% Ca + Na)

Dodavatel: Thermo Fisher Scientific
Varování

36391.14 36391.22
36391.14EA 2850 CZK
36391.14 36391.22
Fluorid hořečnatý ≥99.9% (metals basis excluding Ca, Na) (max. 1% Ca + Na)
Fluorid hořečnatý

Purity: 99.9% CAS No.: 7783-40-6 Molecular Formula: F2Mg Formula Weight: 62.30 Physical Form: 3-6mm Vacuum Melted Lump Density: 3.148 Refractive Index: 1.35-1.38 Appearance: White Odour: Odourless Melting Point: 1261° Merck Reference: 14,5665 MDL No.: MFCD00011108


Vacuum deposition

Vzorec: MgF₂
Bod varu: 2260 °C (1013 hPa)
Bod tání: 1261 °C
Hustota: 3,148
Storage Temperature: Ambient
MDL Number: MFCD00011108
CAS číslo: 7783-40-6
EINECS: 231-995-1
Merck Index: 14,5665

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR