Tisk…

Fluorid vápenatý ≥99.95% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

10998.14 10998.22
10998.14EA 3680 CZK
10998.14 10998.22
Fluorid vápenatý ≥99.95% (metals basis)
Fluorid vápenatý

Purity: 99.95% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: -325 Mesh Powder Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 1403° Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

Vzorec: CaF₂
Bod varu: ∼2500 °C
Bod tání: 1403 °C
Hustota: 3,18
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS číslo: 7789-75-5
EINECS: 232-188-7
Merck Index: 14,1667

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR