Fluorid barnatý, Optical Grade
Dodavatel: Thermo Scientific
|
Varování
|
14057.06
14057.14
14057.22
14057.06EA
1470
CZK
14057.06
14057.14
14057.22
Fluorid barnatý, Optical Grade
Fluorid barnatý
CAS No.: 7787-32-8
Molecular Formula: BaF2
Formula Weight: 175.36
Physical Form: Broken Crystals
Density: 4.83
Refractive Index: 1.4741
Odour: Odourless
Melting Point: 1353°
Boiling Point: 2260°
Merck Reference: 14,974
UN No.: 1564
MDL No.: MFCD00003450
Vzorec:
BaF₂ MW: 175,33 g/mol Bod varu: 2260 °C Bod tání: 1353 °C Hustota: 4,83 Storage Temperature: Ambient |
MDL Number:
MFCD00003450 CAS číslo: 7787-32-8 EINECS: 232-108-0 UN: 1564 ADR: 6.1,III Merck Index: 14,974 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...