Fluorid barnatý ≥99.99% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
10647.14
10647.22
10647.36
10647.88
10647.14EA
4530
CZK
10647.14
10647.22
10647.36
10647.88
Fluorid barnatý ≥99.99% (metals basis), Puratronic®
Fluorid barnatý
Purity: 99.99%
CAS No.: 7787-32-8
Molecular Formula: BaF2
Formula Weight: 175.36
Physical Form: Powder
Density: 4.83
Refractive Index: 1.4741
Odour: Odourless
Melting Point: 1353°
Boiling Point: 2260°
Merck Reference: 14,974
UN No.: 1564
MDL No.: MFCD00003450
Vzorec:
BaF₂ MW: 175.33 Bod varu: 2260 °C Bod tání: 1353 °C Hustota: 4.83 Storage Temperature: Ambient |
MDL Number:
MFCD00003450 CAS číslo: 7787-32-8 EINECS: 232-108-0 UN: 1564 ADR: 6.1 III Merck Index: 14,974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...