Hydroxid-uhličitan hořečnatý hydrate ≥99.996% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
Synonyms:
Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic
10796.06
10796.14
10796.22
10796.06EA
2290
CZK
10796.06
10796.14
10796.22
Hydroxid-uhličitan hořečnatý hydrate ≥99.996% (metals basis), Puratronic®
Hydroxid-uhličitan hořečnatý hydrate
Purity: 99.996%
CAS No.: 39409-82-0
Molecular Formula: MgCO3.Mg(OH)2.xH2O (x≈3)
Formula Weight: 142.63(anhy)
Physical Form: Powder
Appearance: White
Melting Point: dec.
Merck Reference: 14,5660
MDL No.: MFCD00149783
Vzorec:
MgCO₃·Mg(OH)₂·xH₂O MW: 142.63 g/mol (anhy) Bod tání: Dec. Hustota: 2.16 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149783 CAS číslo: 39409-82-0 EINECS: 235-192-7 Merck Index: 14,5660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...