Tisk…

Magnesium carbonate hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Magnesium carbonate hydrate, Magnesium carbonate xhydrate

M-Clarity™ quality level = MQ400

M7179-1KG M7179-500G
SIALM7179-1KGEA 23320 CZK
SIALM7179-1KG SIALM7179-500G
Magnesium carbonate hydrate, Sigma-Aldrich®
Magnesium carbonate hydrate
Vzorec: MgCO₃·xH₂O
MW: 84,31 g/mol (anhy)
Hustota: 3,03 g/cm³ (16 °C)
MDL Number: MFCD00151022
CAS číslo: 23389-33-5
EINECS: 208-915-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR