Tisk…

Barium bromate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11008.22 A11008.36
A11008.22EA 2160 CZK
A11008.22 A11008.36
Barium bromate ≥97%
Barium bromate

Purity: 97% CAS No.: 13967-90-3 Molecular Formula: BaBr2O6 Formula Weight: 393.16 Odour: Odourless UN No.: 2719 MDL No.: MFCD00014177

Vzorec: BaBr₂O₆
Hustota: 3,99
Storage Temperature: Ambient
MDL Number: MFCD00014177
CAS číslo: 13967-90-3
EINECS: 237-750-5
UN: 2719
ADR: 5.1,II
Merck Index: 12,00994

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR