Tisk…

Calcium hexaboride ≥95%

Dodavatel: Thermo Scientific
Varování

41721.18 41721.30 41721.A1
41721.18EA 1680 CZK
41721.18 41721.30 41721.A1
Calcium hexaboride ≥95%
Calcium hexaboride

Purity: 95+% CAS No.: 12007-99-7 Molecular Formula: CaB6 Formula Weight: 104.95 Physical Form: -20 Mesh Powder Density: 2.30 Melting Point: 2235° MDL No.: MFCD00148904

Vzorec: B₆Ca
MW: 104,94 g/mol
Bod tání: 2235 °C
Hustota: 2,30
Storage Temperature: Ambient
MDL Number: MFCD00148904
CAS číslo: 12007-99-7
EINECS: 234-525-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR