Hydrogendifluorid amonný, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Ammonium bifluoride, Ammonium hydrogen fluoride
M-Clarity™ quality level = MQ100
455830-100G
455830-25G
SIAL455830-100GEA
13380
CZK
SIAL455830-100G
SIAL455830-25G
Hydrogendifluorid amonný, Sigma-Aldrich®
Hydrogendifluorid amonný
Vzorec:
NH₄HF₂ MW: 57,04 g/mol Bod tání: 127 °C Hustota: 0,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012018 CAS číslo: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 13,00498 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...