Hydrogendifluorid amonný 95%, Reagent Grade
Dodavatel: Honeywell Chemicals
|
Nebezpečí
|
Synonyms:
Ammonium bifluoride, Ammonium hydrogen fluoride
224820-2.5KG
224820-500G
224820-2.5KGEA
12050
CZK
224820-2.5KG
224820-500G
Hydrogendifluorid amonný 95%, Reagent Grade
Hydrogendifluorid amonný
- ≥95% for general laboratory use.
Vzorec:
NH₄HF₂ MW: 57,04 g/mol Bod tání: 126 °C (Sublimation point 1,013 hPa) Hustota: 1,500 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012018 CAS číslo: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 13,00498 |
Výsledky specifikačního testu
Identity (IR) | complying |
Assay of NH₄HF₂ | 94.0 - 106.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...