Hydrogendifluorid amonný 95% remainder mainly ammonium fluoride
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L17237.22
L17237.36
L17237.0E
L17237.C4
L17237.22EA
982
CZK
L17237.22
L17237.36
L17237.0E
L17237.C4
Hydrogendifluorid amonný 95% remainder mainly ammonium fluoride
Hydrogendifluorid amonný
Purity: 95%
CAS No.: 1341-49-7
Molecular Formula: H5F2N
Formula Weight: 57.04
Odour: Ammonia-like
pH: 2-3
Melting Point: 120-122°
Boiling Point: 238° dec.
Merck Reference: 14,498
UN No.: 1727
MDL No.: MFCD00012018
Vzorec:
NH₄HF₂ Bod varu: 238 °C (rozkl.) Bod tání: 120…122 °C Hustota: 1,500 Storage Temperature: Ambient |
MDL Number:
MFCD00012018 CAS číslo: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 14,498 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...