Tisk…

Hydrogendifluorid amonný ≥95%, tech.

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Ammonium bifluoride, Ammonium hydrogen fluoride

14660.36 14660.A3
14660.36EA 1730 CZK
14660.36 14660.A3
Hydrogendifluorid amonný ≥95%, tech.
Hydrogendifluorid amonný

Purity: 95% CAS No.: 1341-49-7 Molecular Formula: (NH4)HF2 Formula Weight: 57.04 Physical Form: Flake Density: 1.500 pH: 2-3 Melting Point: 120-122° Boiling Point: 238° Merck Reference: 14,498 UN No.: 1727 MDL No.: MFCD00012018

Vzorec: NH₄HF₂
MW: 57,04 g/mol
Bod varu: 238 °C
Bod tání: 120…122 °C
Hustota: 1,500
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS číslo: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 14,498

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR