Sodium tellurite(IV) ≥99% −100 mesh, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Sodium tellurite, Disodium trioxotellurate
M-Clarity™ quality level = MQ100
400688-10G
400688-50G
SIAL400688-10GEA
5120
CZK
SIAL400688-10G
SIAL400688-50G
Sodium tellurite(IV) ≥99% −100 mesh, Sigma-Aldrich®
Sodium tellurite(IV)
Vzorec:
Na₂O₃Te MW: 221,58 g/mol Bod tání: 710 °C |
MDL Number:
MFCD00036141 CAS číslo: 10102-20-2 EINECS: 233-268-4 UN: 3284 ADR: 6.1,II Merck Index: 12,08834 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...