Tisk…

Lithium tantalate ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

41834.09 41834.18
41834.09EA 4770 CZK
41834.09 41834.18
Lithium tantalate ≥99.9% (metals basis)
Lithium tantalate

Purity: 99.9% CAS No.: 12031-66-2 Molecular Formula: LiTaO3 Formula Weight: 235.89 Physical Form: -200 Mesh Powder Odour: Odourless MDL No.: MFCD00016174

Vzorec: LiO₃Ta
Storage Temperature: Ambient
MDL Number: MFCD00016174
CAS číslo: 12031-66-2
EINECS: 234-757-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR