Tisk…

Hydrogensíran draselný ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Potassium bisulphate

A12428.0C A12428.0E A12428.22 A12428.36
A12428.0CEA 15310 CZK
A12428.0C A12428.0E A12428.22 A12428.36
Hydrogensíran draselný ≥97%
Hydrogensíran draselný

Purity: 97% CAS No.: 7646-93-7 Molecular Formula: HKO4S Formula Weight: 136.17 Odour: Odourless Melting Point: 195-197° dec. UN No.: 2509 MDL No.: MFCD00011404

Vzorec: KHSO₄
MW: 136.17
Bod tání: 195…197 °C (rozkl.)
Hustota: 2.240
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS číslo: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR