Tisk…

Síran draselný ≥99.0% ACS

Dodavatel: Thermo Fisher Scientific

14311.36 14311.A3
14311.36EA 1430 CZK
14311.36 14311.A3
Síran draselný ≥99.0% ACS
Síran draselný

Purity: 99.0% min CAS No.: 7778-80-5 Molecular Formula: K2SO4 Formula Weight: 174.27 Physical Form: Crystalline Density: 2.662 Melting Point: 1069° Boiling Point: 1689° Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,7674 MDL No.: MFCD00011388

Vzorec: K₂SO₄
Bod varu: 1689 °C
Bod tání: 1069 °C
Hustota: 2,662
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS číslo: 7778-80-5
EINECS: 231-915-5
Merck Index: 14,7674

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR