Sodium hexafluoroantimonate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
237981-100G
237981-25G
SIAL237981-100GEA
5450
CZK
SIAL237981-100G
SIAL237981-25G
Sodium hexafluoroantimonate, Sigma-Aldrich®
Sodium hexafluoroantimonate
Vzorec:
F₆NaSb Hustota: 3,379 g/cm³ (18 °C) |
MDL Number:
MFCD00003482 CAS číslo: 16925-25-0 EINECS: 240-989-8 UN: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...