Tisk…

Disíran didraselný ≥99%, ReagentPlus®, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Potassium disulphate (Potassium pyrosulphate), Dipotassium disulphate, Potassium pyrosulphate

M-Clarity™ quality level = MQ100

307750-25G 307750-500G
SIAL307750-25GEA 1610 CZK
SIAL307750-25G SIAL307750-500G
Disíran didraselný ≥99%, ReagentPlus®, Sigma-Aldrich®
Disíran didraselný
Vzorec: K₂O₇S₂
MW: 254,32 g/mol
Bod tání: ∼325 °C
Hustota: 2,51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011385
CAS číslo: 7790-62-7
EINECS: 232-216-8
UN: 2923
ADR: 8 (6.1),I
Merck Index: 13,07749

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR