Tisk…

Hexachlorplatičitan didraselný (39,6% Pt)

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Dipotassium hexachloroplatinate

12169.06 12169.01 12169.03
12169.06EA 14380 CZK
12169.06 12169.01 12169.03
Hexachlorplatičitan didraselný (39,6% Pt)
Hexachlorplatičitan didraselný

CAS No.: 16921-30-5 Molecular Formula: Cl6K2Pt Formula Weight: 486.01 Physical Form: Crystalline powder Density: 3.50 Refractive Index: ≈1.825 Sensitivity: Hygroscopic Appearance: Yellow Odour: Odourless Melting Point: 250°dec. Solubility: Soluble in hot water. Slightly soluble in cold water. Practically insoluble in alcohol Merck Reference: 14,7636 UN No.: 3288 MDL No.: MFCD00011389

Vzorec: K₂PtCl₆
MW: 485,99 g/mol
Bod tání: 250 °C, (rozkl.)
Hustota: 3,50
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS číslo: 16921-30-5
EINECS: 240-979-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,7636

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR