Tetrachlorplatnatan draselný ≥99.99% (metals basis) Pt ≥ 46.0%, Premion®
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate
43946.03
43946.06
43946.14
43946.03EA
5940
CZK
43946.03
43946.06
43946.14
Tetrachlorplatnatan draselný ≥99.99% (metals basis) Pt ≥ 46.0%, Premion®
Tetrachlorplatnatan draselný
Purity: 99.99%
CAS No.: 10025-99-7
Molecular Formula: Cl4K2Pt
Formula Weight: 415.11
Physical Form: Crystalline
Density: 3.38
Appearance: Red-brown
Odour: Odourless
Melting Point: dec.
Merck Reference: 14,7682
UN No.: 3288
MDL No.: MFCD00011378
Vzorec:
Cl₄K₂Pt MW: 415,09 g/mol Bod varu: 35…40 °C (20 mmHg) Bod tání: Dec. Hustota: 3,38 Storage Temperature: Ambient |
MDL Number:
MFCD00011378 CAS číslo: 10025-99-7 EINECS: 233-050-9 UN: 3288 ADR: 6.1,III Merck Index: 14,7682 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...