Tisk…

Tetrachlorplatnatan draselný (≥46,0% Pt) ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate

11048.03 11048.06 11048.14
11048.03EA 4260 CZK
11048.03 11048.06 11048.14
Tetrachlorplatnatan draselný (≥46,0% Pt) ≥99.9% (metals basis)
Tetrachlorplatnatan draselný

Purity: 99.9% CAS No.: 10025-99-7 Molecular Formula: Cl4K2Pt Formula Weight: 415.11 Physical Form: Crystalline Density: 3.38 Appearance: Red-brown Odour: Odourless Melting Point: dec. Solubility: Soluble in water Merck Reference: 14,7682 UN No.: 3288 MDL No.: MFCD00011378

Vzorec: Cl₄K₂Pt
Bod varu: 35…40 °C (20 mmHg)
Bod tání: Dec.
Hustota: 3,38
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS číslo: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III
Merck Index: 14,7682

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR